3-(Benzyloxy)benzoyl chloride 95%

Supplier: Thermo Scientific
Danger

H50410.03 H50410.MD
ALFAH50410.03EA 562 SGD
ALFAH50410.03 ALFAH50410.MD
3-(Benzyloxy)benzoyl chloride 95%
3-(Benzyloxy)benzoyl chloride

Purity: 95% CAS No.: 61535-46-4 Molecular Formula: C14H11ClO2 Formula Weight: 246.70 Melting Point: 40-42° UN No.: UN3261 MDL No.: MFCD02258040

Formula: C₁₄H₁₁ClO₂
MW: 246,7 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02258040
CAS Number: 61535-46-4
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR