2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde 99%

Supplier: Thermo Scientific
Warning

H50466.03 H50466.MD
ALFAH50466.03EA 527 SGD
ALFAH50466.03 ALFAH50466.MD
2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde 99%
2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde

Purity: 99% CAS No.: 320423-50-5 Molecular Formula: C9H13N3OS Formula Weight: 211.29 MDL No.: MFCD01568852

Formula: C₉H₁₃N₃OS
Storage Temperature: Ambient
MDL Number: MFCD01568852
CAS Number: 320423-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR