6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

Supplier: Thermo Scientific

H50585.03 H50585.MD
ALFAH50585.03EA 482 SGD
ALFAH50585.03 ALFAH50585.MD
6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one
6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

CAS No.: 19165-25-4 Molecular Formula: C9H6BrNO2 Formula Weight: 240.06 MDL No.: MFCD00115891

Formula: C₉H₆BrNO₂
MW: 240,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115891
CAS Number: 19165-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR