7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole 99%

Supplier: Thermo Scientific

Synonyms: 7-Bromo-1,2,3,4-tetrahydrocyclopenta[b]indole

H50592.03 H50592.MD
ALFAH50592.03EA 527 SGD
ALFAH50592.03 ALFAH50592.MD
7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole 99%
7-Bromo-1,2,3,4-tetrahydrocyclopent[b]indole

Purity: 99% CAS No.: 164736-47-4 Molecular Formula: C11H10BrN Formula Weight: 236.11 MDL No.: MFCD00239009

Formula: C₁₁H₁₀BrN
MW: 236,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239009
CAS Number: 164736-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR