N-(2-Bromobenzyl)acetamide 97%

Supplier: Thermo Scientific
Warning

H50599.03 H50599.06
ALFAH50599.03EA 296 SGD
ALFAH50599.03 ALFAH50599.06
N-(2-Bromobenzyl)acetamide 97%
N-(2-Bromobenzyl)acetamide

Purity: 97% CAS No.: 74315-07-4 Molecular Formula: C9H10BrNO Formula Weight: 228.09 Melting Point: 78-80° MDL No.: MFCD00831169

Formula: C₉H₁₀BrNO
MW: 228,09 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00831169
CAS Number: 74315-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR