2-(2-Chlorophenoxy)ethylamine 95%
Supplier: Thermo Scientific
|
Danger
|
H50704.03
H50704.MD
ALFAH50704.03EA
466
SGD
ALFAH50704.03
ALFAH50704.MD
2-(2-Chlorophenoxy)ethylamine 95%
2-(2-Chlorophenoxy)ethylamine
Purity: 95%
CAS No.: 26378-53-0
Molecular Formula: C8H10ClNO
Formula Weight: 171.63
Melting Point: 39-40°
Boiling Point: 150-153°/18mm
UN No.: UN2735
MDL No.: MFCD00125294
Formula:
C₈H₁₀ClNO MW: 171,63 g/mol Boiling Pt: 150…153 °C (18 mmHg) Melting Pt: 39…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00125294 CAS Number: 26378-53-0 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...