3-(1-Piperazino)propionitrile

Supplier: Thermo Scientific
Danger

H50710.03 H50710.06
ALFAH50710.03EA 133 SGD
ALFAH50710.03 ALFAH50710.06
3-(1-Piperazino)propionitrile
3-(1-Piperazino)propionitrile

CAS No.: 34064-86-3 Molecular Formula: C7H13N3 Formula Weight: 139.20 UN No.: UN3276 MDL No.: MFCD02093566

Formula: C₇H₁₃N₃
MW: 139,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093566
CAS Number: 34064-86-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR