1-(4-Methoxyphenyl)-2-methylaminoethanol 97%

Supplier: Thermo Scientific
Warning

H50754.03 H50754.MD
ALFAH50754.03EA 483 SGD
ALFAH50754.03 ALFAH50754.MD
1-(4-Methoxyphenyl)-2-methylaminoethanol 97%
1-(4-Methoxyphenyl)-2-methylaminoethanol

Purity: 97% CAS No.: 58777-87-0 Molecular Formula: C10H15NO2 Formula Weight: 181.24 MDL No.: MFCD08060969

Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD08060969
CAS Number: 58777-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR