4-Amino-1-(3-methyl-2-pyridyl)piperidine 99%
Supplier: Thermo Scientific
|
Warning
|
H50779.03
H50779.MD
ALFAH50779.03EA
614
SGD
ALFAH50779.03
ALFAH50779.MD
4-Amino-1-(3-methyl-2-pyridyl)piperidine 99%
4-Amino-1-(3-methyl-2-pyridyl)piperidine
Purity: 99%
CAS No.: 902837-09-6
Molecular Formula: C11H17N3
Formula Weight: 191.28
MDL No.: MFCD08060981
Formula:
C₁₁H₁₇N₃ MW: 191,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08060981 CAS Number: 902837-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...