4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one 96%
Supplier: Thermo Scientific
|
Warning
|
H50828.03
H50828.MD
ALFAH50828.03EA
0
SGD
ALFAH50828.03
ALFAH50828.MD
4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one 96%
4-Phenyl-1-piperidin-4-yl-1,3-dihydro-2H-imidazol-2-one
Purity: 96%
CAS No.: 205058-28-2
Molecular Formula: C14H17N3O
Formula Weight: 243.31
Melting Point: ca 235° dec.
MDL No.: MFCD08061015
Formula:
C₁₄H₁₇N₃O Melting Pt: ∼ 235 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD08061015 CAS Number: 205058-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...