3-Methyl-2-(1-piperazinyl)quinoline 97%

Supplier: Thermo Scientific
Warning

H50834.03 H50834.MD
ALFAH50834.03EA 0 SGD
ALFAH50834.03 ALFAH50834.MD
3-Methyl-2-(1-piperazinyl)quinoline 97%
3-Methyl-2-(1-piperazinyl)quinoline

Purity: 97% CAS No.: 348133-74-4 Molecular Formula: C14H17N3 Formula Weight: 227.31 MDL No.: MFCD08061017

Formula: C₁₄H₁₇N₃
MW: 227,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061017
CAS Number: 348133-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR