1-(2-Aminoethyl)-4-methylhomopiperazine 98%

Supplier: Thermo Scientific
Danger

H50837.03 H50837.MD
ALFAH50837.03EA 572 SGD
ALFAH50837.03 ALFAH50837.MD
1-(2-Aminoethyl)-4-methylhomopiperazine 98%
1-(2-Aminoethyl)-4-methylhomopiperazine

Purity: 98% CAS No.: 39270-45-6 Molecular Formula: C8H19N3 Formula Weight: 157.26 UN No.: UN2735 MDL No.: MFCD08061018

Formula: C₈H₁₉N₃
MW: 157,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061018
CAS Number: 39270-45-6
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR