4-(Methoxymethyl)phenylacetic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H50848.03
H50848.MD
ALFAH50848.03EA
476
SGD
ALFAH50848.03
ALFAH50848.MD
4-(Methoxymethyl)phenylacetic acid 97%
4-(Methoxymethyl)phenylacetic acid
Purity: 97%
CAS No.: 343880-24-0
Molecular Formula: C10H12O3
Formula Weight: 180.21
MDL No.: MFCD08060493
Formula:
C₁₀H₁₂O₃ MW: 180,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08060493 CAS Number: 343880-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...