N-Ethyl-2-(1-piperazinyl)nicotinamide 99%
Supplier: Thermo Scientific
|
Warning
|
H50856.03
H50856.MD
ALFAH50856.03EA
500
SGD
ALFAH50856.03
ALFAH50856.MD
N-Ethyl-2-(1-piperazinyl)nicotinamide 99%
N-Ethyl-2-(1-piperazinyl)nicotinamide
Purity: 99%
Molecular Formula: C12H18N4O
Formula Weight: 234.30
MDL No.: MFCD08061031
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...