3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine 97%

Supplier: Thermo Scientific
Warning

H50879.03 H50879.MD
ALFAH50879.03EA 505 SGD
ALFAH50879.03 ALFAH50879.MD
3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine 97%
3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine

Purity: 97% CAS No.: 139269-06-0 Molecular Formula: C9H15N3O Formula Weight: 181.24 MDL No.: MFCD08061044

Formula: C₉H₁₅N₃O
MW: 181,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061044
CAS Number: 139269-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR