3-Amino-5-cyclobutyl-1H-pyrazole 97%

Supplier: Thermo Scientific
Warning

H50880.03 H50880.MD
ALFAH50880.03EA 514 SGD
ALFAH50880.03 ALFAH50880.MD
3-Amino-5-cyclobutyl-1H-pyrazole 97%
3-Amino-5-cyclobutyl-1H-pyrazole

Purity: 97% CAS No.: 326827-21-8 Molecular Formula: C7H11N3 Formula Weight: 137.19 MDL No.: MFCD08061045

Formula: C₇H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD08061045
CAS Number: 326827-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR