3-(2-Methoxyphenoxy)pyrrolidine 98%

Supplier: Thermo Scientific
Warning

H50883.03 H50883.MD
ALFAH50883.03EA 517 SGD
ALFAH50883.03 ALFAH50883.MD
3-(2-Methoxyphenoxy)pyrrolidine 98%
3-(2-Methoxyphenoxy)pyrrolidine

Purity: 98% CAS No.: 21767-12-4 Molecular Formula: C8H13N3O Formula Weight: 167.21 MDL No.: MFCD07801211

Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD08060986
CAS Number: 21767-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR