1-(5-Methyl-2-pyridinyl)piperazine 97%

Supplier: Thermo Scientific
Warning

H50897.03 H50897.MD
ALFAH50897.03EA 448 SGD
ALFAH50897.03 ALFAH50897.MD
1-(5-Methyl-2-pyridinyl)piperazine 97%
1-(5-Methyl-2-pyridinyl)piperazine

Purity: 97% CAS No.: 104395-86-0 Molecular Formula: C10H15N3 Formula Weight: 177.25 MDL No.: MFCD08061050

Formula: C₁₀H₁₅N₃
MW: 177,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061050
CAS Number: 104395-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR