3-Benzyloxyphenylacetic acid 95%
Supplier: Thermo Scientific
|
Warning
|
H50908.03
H50908.MD
ALFAH50908.03EA
380
SGD
ALFAH50908.03
ALFAH50908.MD
3-Benzyloxyphenylacetic acid 95%
3-Benzyloxyphenylacetic acid
Purity: 95%
CAS No.: 1860-58-8
Molecular Formula: C15H14O3
Formula Weight: 242.28
MDL No.: MFCD02664806
Formula:
C₁₅H₁₄O₃ MW: 242,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664806 CAS Number: 1860-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...