4-Amino-1-isobutyrylpiperidine 95%

Supplier: Thermo Scientific
Danger

H50916.03 H50916.MD
ALFAH50916.03EA 479 SGD
ALFAH50916.03 ALFAH50916.MD
4-Amino-1-isobutyrylpiperidine 95%
4-Amino-1-isobutyrylpiperidine

Purity: 95% CAS No.: 172478-05-6 Molecular Formula: C9H18N2O Formula Weight: 170.26 MDL No.: MFCD08061057

Formula: C₉H₁₈N₂O
MW: 170,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061057
CAS Number: 172478-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR