4-Aminomethyl-1-(n-butyl)piperidine 97%
Supplier: Thermo Scientific
|
Warning
|
H50947.03
H50947.MD
ALFAH50947.03EA
410
SGD
ALFAH50947.03
ALFAH50947.MD
4-Aminomethyl-1-(n-butyl)piperidine 97%
4-Aminomethyl-1-(n-butyl)piperidine
Purity: 97%
CAS No.: 65017-57-4
Molecular Formula: C10H22N2
Formula Weight: 170.30
MDL No.: MFCD08060490
Formula:
C₁₀H₂₂N₂ MW: 170,30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08060490 CAS Number: 65017-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...