5-Propyl-1H-pyrazol-3-ylamine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Propyl-1H-pyrazol-3-amine

H50969.03 H50969.MD
ALFAH50969.03EA 541 SGD
ALFAH50969.03 ALFAH50969.MD
5-Propyl-1H-pyrazol-3-ylamine 97%
5-Propyl-1H-pyrazol-3-ylamine

Purity: 97% CAS No.: 126748-58-1 Molecular Formula: C6H11N3 Formula Weight: 125.17 MDL No.: MFCD08061095

Formula: C₆H₁₁N₃
MW: 125,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061095
CAS Number: 126748-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR