4-(Ethoxymethyl)piperidine 98%

Supplier: Thermo Scientific
Warning

H50975.03 H50975.MD
ALFAH50975.03EA 499 SGD
ALFAH50975.03 ALFAH50975.MD
4-(Ethoxymethyl)piperidine 98%
4-(Ethoxymethyl)piperidine

Purity: 98% CAS No.: 265108-38-1 Molecular Formula: C8H17NO Formula Weight: 143.23 MDL No.: MFCD06247374

Formula: C₈H₁₇NO
MW: 143,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247374
CAS Number: 265108-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR