4-Acetamidopiperidine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-(Piperidin-4-yl)acetamide
H50981.03
H50981.MD
ALFAH50981.03EA
286
SGD
ALFAH50981.03
ALFAH50981.MD
4-Acetamidopiperidine 97%
4-Acetamidopiperidine
Purity: 97%
CAS No.: 5810-56-0
Molecular Formula: C7H14N2O
Formula Weight: 142.20
Melting Point: 141°
MDL No.: MFCD05254805
Formula:
C₇H₁₄N₂O MW: 142,20 g/mol Melting Pt: 141 °C Storage Temperature: Ambient |
MDL Number:
MFCD05254805 CAS Number: 5810-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...