2-(4-Fluorophenyl)-2-methylmorpholine 99%

Supplier: Thermo Scientific
Warning

H51007.03 H51007.MD
ALFAH51007.03EA 0 SGD
ALFAH51007.03 ALFAH51007.MD
2-(4-Fluorophenyl)-2-methylmorpholine 99%
2-(4-Fluorophenyl)-2-methylmorpholine

CAS No.: 109461-46-3 Molecular Formula: C11H14FNO Formula Weight: 195.24 MDL No.: MFCD08061112

Formula: C₁₁H₁₄FNO
MW: 195,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061112
CAS Number: 109461-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR