4-(Phenoxymethyl)phenylacetic acid 99%
Supplier: Thermo Scientific
|
Warning
|
H51008.03
H51008.MD
ALFAH51008.03EA
493
SGD
ALFAH51008.03
ALFAH51008.MD
4-(Phenoxymethyl)phenylacetic acid 99%
4-(Phenoxymethyl)phenylacetic acid
Purity: 99%
CAS No.: 344349-71-9
Molecular Formula: C15H14O3
Formula Weight: 242.28
MDL No.: MFCD08061113
Formula:
C₁₅H₁₄O₃ MW: 242,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061113 CAS Number: 344349-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...