2-(4-Chlorophenyl)-2-methylmorpholine 99%

Supplier: Thermo Scientific
Warning

H51010.03 H51010.MD
ALFAH51010.03EA 0 SGD
ALFAH51010.03 ALFAH51010.MD
2-(4-Chlorophenyl)-2-methylmorpholine 99%
2-(4-Chlorophenyl)-2-methylmorpholine

Purity: 99% CAS No.: 109461-44-1 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD08061115

Formula: C₁₁H₁₄ClNO
MW: 211,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061115
CAS Number: 109461-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR