α-(4-tert-Butylphenyl)di(2-pyrrolyl)methane 99%

Supplier: Thermo Scientific
Warning

H51066.03 H51066.06
ALFAH51066.03EA 236 SGD
ALFAH51066.03 ALFAH51066.06
α-(4-tert-Butylphenyl)di(2-pyrrolyl)methane 99%
alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane

Purity: 99% CAS No.: 167482-98-6 Molecular Formula: C19H22N2 Formula Weight: 278.39 Melting Point: 158-162° MDL No.: MFCD03454220

Formula: C₁₉H₂₂N₂
MW: 278,39 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03454220
CAS Number: 167482-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR