6-Chloro-7-iodo-7-deazapurine 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine

H51128.03 H51128.06
ALFAH51128.03EA 250 SGD
ALFAH51128.03 ALFAH51128.06
6-Chloro-7-iodo-7-deazapurine 96%
6-Chloro-7-iodo-7-deazapurine

Purity: 96% CAS No.: 123148-78-7 Molecular Formula: C6H3ClIN3 Formula Weight: 279.47 Melting Point: ca 180° dec. MDL No.: MFCD09263258

Formula: C₆H₃ClIN₃
MW: 279.47 g/mol
Melting Pt: ∼ 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09263258
CAS Number: 123148-78-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR