4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Cyano-3-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-acrylic acid ethyl ester
H51130.03
H51130.14
ALFAH51130.03EA
53.8
SGD
ALFAH51130.03
ALFAH51130.14
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester 97%
4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
Molecular Formula: C18H22BNO4
Formula Weight: 327.18
MDL No.: MFCD11113044
Formula:
C₁₈H₂₂BNO₄ MW: 327,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11113044 CAS Number: 1218790-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...