4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester

Supplier: Thermo Scientific
Warning

H51671.03 H51671.06
ALFAH51671.03EA 116 SGD
ALFAH51671.03 ALFAH51671.06
4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester
4-[3-(1-Imidazolyl)propylaminomethyl]benzeneboronic acid pinacol ester

CAS No.: 1257648-78-4 Molecular Formula: C19H28BN3O2 Formula Weight: 341.26 MDL No.: MFCD12546046

Formula: C₁₉H₂₈BN₃O₂
MW: 341,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546046
CAS Number: 1257648-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR