N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride 96%
Supplier: Thermo Scientific
H51679.MD
H51679.03
ALFAH51679.MDEA
171
SGD
ALFAH51679.MD
ALFAH51679.03
N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride 96%
N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
Purity: 96%
CAS No.: 902836-08-2
Molecular Formula: C12H18N4O.HCl
Formula Weight: 270.76
MDL No.: MFCD12407084
Formula:
C₁₂H₁₈N₄O·HCl MW: 270,76 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12407084 CAS Number: 902836-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...