2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(3-Fluorophenoxy)ethylamine
H51681.MD
H51681.03
ALFAH51681.MDEA
138
SGD
ALFAH51681.MD
ALFAH51681.03
2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
2-(3-Fluorophenoxy)ethylamine hydrochloride
Purity: 98%
CAS No.: 120351-93-1
Molecular Formula: C8H10FNO.HCl
Formula Weight: 191.63
MDL No.: MFCD11099553
Formula:
C₈H₁₁ClFNO MW: 191,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11099553 CAS Number: 120351-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...