Benzyl thioacetimidate hydrochloride 96%

Supplier: Thermo Scientific
Danger

H51844.03 H51844.06 H51844.14
ALFAH51844.03EA 53.1 SGD
ALFAH51844.03 ALFAH51844.06 ALFAH51844.14
Benzyl thioacetimidate hydrochloride 96%
Benzyl thioacetimidate hydrochloride

Purity: 96% CAS No.: 32894-07-8 Molecular Formula: C9H11NS.HCl Formula Weight: 201.71 MDL No.: MFCD05865210

Formula: C₉H₁₁NS·HCl
MW: 201,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865210
CAS Number: 32894-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR