4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H51850.MD
H51850.03
ALFAH51850.MDEA
55.9
SGD
ALFAH51850.MD
ALFAH51850.03
4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid 97%
4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid
Purity: 97%
Molecular Formula: C15H10N2O2S
Formula Weight: 282.32
MDL No.: MFCD16251584
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...