2-Chloro-1-(ethoxymethyl)imidazole 97%
Supplier: Thermo Scientific
|
Warning
|
H51870.MD
H51870.03
ALFAH51870.MDEA
0
SGD
ALFAH51870.MD
ALFAH51870.03
2-Chloro-1-(ethoxymethyl)imidazole 97%
2-Chloro-1-(ethoxymethyl)imidazole
Purity: 97%
CAS No.: 850429-55-9
Molecular Formula: C6H9ClN2O
Formula Weight: 160.60
MDL No.: MFCD03788569
Formula:
C₆H₉ClN₂O MW: 160,60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788569 CAS Number: 850429-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...