2-Chloro-4-fluoro-3-methylbenzoic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H51879.MD
H51879.03
ALFAH51879.MDEA
213
SGD
ALFAH51879.MD
ALFAH51879.03
2-Chloro-4-fluoro-3-methylbenzoic acid 97%
2-Chloro-4-fluoro-3-methylbenzoic acid
Purity: 97%
Molecular Formula: C8H6ClFO2
Formula Weight: 188.59
Melting Point: 186-189°
MDL No.: MFCD11846444
Formula:
C₈H₆ClFO₂ MW: 188,59 g/mol Melting Pt: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD11846444 CAS Number: 173315-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...