3-Ethynylbenzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(3-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(3-Ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H51902.03 H51902.06
ALFAH51902.03EA 220 SGD
ALFAH51902.03 ALFAH51902.06
3-Ethynylbenzeneboronic acid pinacol ester 95%
3-Ethynylbenzeneboronic acid pinacol ester

Purity: 95% Molecular Formula: C14H17BO2 Formula Weight: 228.10 MDL No.: MFCD16294529

Formula: C₁₄H₁₇BO₂
MW: 228,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294529
CAS Number: 946168-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR