4-(1-Piperidinyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific

Synonyms: 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-piperidine

H51922.03 H51922.06
ALFAH51922.03EA 162 SGD
ALFAH51922.03 ALFAH51922.06
4-(1-Piperidinyl)benzeneboronic acid pinacol ester 95%
4-(1-Piperidinyl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 852227-96-4 Molecular Formula: C17H26BNO2 Formula Weight: 287.21 MDL No.: MFCD07368524

Formula: C₁₇H₂₆BNO₂
MW: 287,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368524
CAS Number: 852227-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR