9-Benzylcarbazole-3-boronic acid pinacol ester 95%

Supplier: Thermo Scientific

Synonyms: 9-Benzyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-9H-carbazole

H51944.03 H51944.06
ALFAH51944.03EA 93.1 SGD
ALFAH51944.03 ALFAH51944.06
9-Benzylcarbazole-3-boronic acid pinacol ester 95%
9-Benzylcarbazole-3-boronic acid pinacol ester

Purity: 95% Molecular Formula: C25H26BNO2 Formula Weight: 383.30 MDL No.: MFCD16294547

Formula: C₂₅H₂₆BNO₂
MW: 383,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294547
CAS Number: 1357387-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR