1-Benzyl-7-azaindole-3-boronic acid pinacol ester 95%

Supplier: Thermo Scientific

Synonyms: 1-Benzyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine, 1-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

H51949.03 H51949.06
ALFAH51949.03EA 120 SGD
ALFAH51949.03 ALFAH51949.06
1-Benzyl-7-azaindole-3-boronic acid pinacol ester 95%
1-Benzyl-7-azaindole-3-boronic acid pinacol ester

Purity: 95% Molecular Formula: C20H23BN2O2 Formula Weight: 334.22 MDL No.: MFCD16294550

Formula: C₂₀H₂₃BN₂O₂
MW: 334,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294550
CAS Number: 1350626-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR