9,9-Di-n-octylfluorene-2-boronic acid pinacol ester 95%
Supplier: Thermo Scientific
Synonyms:
9,9-Di-n-octylfluorene-2-boronic acid Pinacol ester, 9,9-Di-n-octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene, 2-(9,9-Di-n-octyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H51953.03
H51953.06
ALFAH51953.03EA
109
SGD
ALFAH51953.03
ALFAH51953.06
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester 95%
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
Purity: 95%
Molecular Formula: C35H53BO2
Formula Weight: 516.61
MDL No.: MFCD16294553
Formula:
C₃₅H₅₃BO₂ MW: 516,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294553 CAS Number: 302554-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...