Fmoc-L-homophenylalanine 95%

Supplier: Thermo Scientific

Synonyms: Fmoc-HoPhe-OH, (S)-2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-phenylbutyric acid, Fmoc-Homophe-OH, N-Fmoc-L-homophenylalanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine, (S)-2-(Fmoc-amino)-4-phenylbutyric acid

H51973.MD H51973.03 H51973.06
ALFAH51973.MDEA 73.3 SGD
ALFAH51973.MD ALFAH51973.03 ALFAH51973.06
Fmoc-L-homophenylalanine 95%
Fmoc-L-homophenylalanine

Purity: 95% CAS No.: 132684-59-4 Molecular Formula: C25H23NO4 Formula Weight: 401.46 Beilstein Reference: 4847669 MDL No.: MFCD00077046

Formula: C₂₅H₂₃NO₄
MW: 401,462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077046
CAS Number: 132684-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR