2-Benzyl-D-proline hydrochloride 95%
Supplier: Thermo Scientific
H52019.MD
H52019.03
ALFAH52019.MDEA
0
SGD
ALFAH52019.MD
ALFAH52019.03
2-Benzyl-D-proline hydrochloride 95%
2-Benzyl-D-proline hydrochloride
Purity: 95%
CAS No.: 637020-57-6
Molecular Formula: C12H15NO2.HCl
Formula Weight: 241.72
Optical Rotation: +33° (c=1 in 1N HCl)
MDL No.: MFCD07367715
Formula:
C₁₂H₁₅NO₂·HCl MW: 241,72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07367715 CAS Number: 637020-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...