trans-N-Boc-4-(2-naphthylmethyl)-L-proline 95%

Supplier: Thermo Scientific

H52035.MD H52035.03
ALFAH52035.MDEA 410 SGD
ALFAH52035.MD ALFAH52035.03
trans-N-Boc-4-(2-naphthylmethyl)-L-proline 95%
trans-N-Boc-4-(2-naphthylmethyl)-L-proline

Purity: 95% Molecular Formula: C21H25NO4 Formula Weight: 355.43 MDL No.: MFCD16658816

Formula: C₂₁H₂₅NO₄
MW: 355,43 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD16658816
CAS Number: 959573-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR