Fmoc-L-β-homophenylalanine 95%

Supplier: Thermo Scientific

Synonyms: Fmoc-B-HoPhe-OH, Fmoc-β-HoPhe-OH, Fmoc-β-HomoPhe-OH

H52052.MD H52052.03
ALFAH52052.MDEA 0 SGD
ALFAH52052.MD ALFAH52052.03
Fmoc-L-β-homophenylalanine 95%
Fmoc-L-beta-homophenylalanine

Purity: 95% CAS No.: 193954-28-8 Molecular Formula: C25H23NO4 Formula Weight: 401.46 Beilstein Reference: 7726557 MDL No.: MFCD01863055

Formula: C₂₅H₂₃NO₄
MW: 401,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863055
CAS Number: 193954-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR