Boc-L-allylglycine . DCHA 95%

Supplier: Thermo Scientific

Synonyms: Boc-Allyl-Gly-OH·DCHA, Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)pent-4-enoate, (S)-N-BOC-alpha-Allylglycine dicyclohexylamine salt

H52062.MD H52062.03 H52062.06
ALFAH52062.MDEA 0 SGD
ALFAH52062.MD ALFAH52062.03 ALFAH52062.06
Boc-L-allylglycine . DCHA 95%
Boc-L-allylglycine . DCHA

Purity: 95% CAS No.: 143979-15-1 Molecular Formula: C10H17NO4.C12H23N Formula Weight: 396.57 MDL No.: MFCD01321013

Formula: C₂₂H₄₀N₂O₄
MW: 396,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321013
CAS Number: 143979-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR