Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid 95%

Supplier: Thermo Scientific

Synonyms: (R)-4-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid

H52136.MD H52136.03
ALFAH52136.MDEA 0 SGD
ALFAH52136.MD ALFAH52136.03
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid 95%
Boc-(R)-3-amino-4-(4-bromophenyl)butyric acid

Purity: 95% CAS No.: 331763-75-8 Molecular Formula: C15H20BrNO4 Formula Weight: 358.23 Melting Point: 144-146° MDL No.: MFCD01860917

Formula: C₁₅H₂₀BrNO₄
MW: 358,24 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD01860917
CAS Number: 331763-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR