2-Methyl-4-(4-pyridyl)thiazole 97%
Supplier: Thermo Scientific
|
Warning
|
H52155.03
H52155.06
ALFAH52155.03EA
201
SGD
ALFAH52155.03
ALFAH52155.06
2-Methyl-4-(4-pyridyl)thiazole 97%
2-Methyl-4-(4-pyridyl)thiazole
Purity: 97%
Molecular Formula: C9H8N2S
Formula Weight: 176.24
MDL No.: MFCD16658845
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...