3,3-Diphenyl-L-alanine 95%

Supplier: Thermo Scientific
Danger

Synonyms: H-Ala(3,3-DiPhenyl)-OH, (S)-2-Amino-3,3-diphenylpropanoic acid

H52169.MD H52169.03 H52169.06
ALFAH52169.MDEA 169 SGD
ALFAH52169.MD ALFAH52169.03 ALFAH52169.06
3,3-Diphenyl-L-alanine 95%
3,3-Diphenyl-L-alanine

Purity: 95% CAS No.: 149597-92-2 Molecular Formula: C15H15NO2 Formula Weight: 241.29 UN No.: UN2811 MDL No.: MFCD01631989

Formula: C₁₅H₁₅NO₂
MW: 241,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631989
CAS Number: 149597-92-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR